Search
Search

RPAP3| Gene Symbol: RPAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005611105.3 | XP_005611162.2 | 2043 | 81 | ||
| XM_005611106.3 | XP_005611163.2 | 1983 | 81 | |||
| XM_005611107.3 | XP_005611164.2 | 2008 | 81 | |||
| GenBank mRNA | JL619824.1 | - | 1 - 2 | 628 | 81 | |
Set Membership: |
|