Search
Search

RC3H1| Gene Symbol: RC3H1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023640576.1 | XP_023496344.1 | 3210 | 104 | ||
| XM_023640577.1 | XP_023496345.1 | 3206 | 104 | |||
| XM_023640578.1 | XP_023496346.1 | 3208 | 104 | |||
| XM_023640579.1 | XP_023496347.1 | 3059 | 104 | |||
| XM_023640581.1 | XP_023496349.1 | 3212 | 104 | |||
Set Membership: |
|