Search
Search

C1H10orf90| Gene Symbol: C1H10orf90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001488777.5 | XP_001488827.3 | 1697 | 101 | ||
| XM_023637137.1 | XP_023492905.1 | 2399 | 101 | |||
| XM_023637146.1 | XP_023492914.1 | 2397 | 101 | |||
| XM_023637148.1 | XP_023492916.1 | 3910 | 101 | |||
| XM_023637153.1 | XP_023492921.1 | 3281 | 101 | |||
| XM_023637162.1 | XP_023492930.1 | 2121 | 101 | |||
Set Membership: |
|