Search
Search

CFAP70| Gene Symbol: CFAP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503856.4 | XP_001503906.1 | 3397 | 116 | ||
| XM_023645010.1 | XP_023500778.1 | 3416 | 116 | |||
| XM_023645015.1 | XP_023500783.1 | 3619 | 116 | |||
| XM_023645023.1 | XP_023500791.1 | 3516 | 116 | |||
| XM_023645035.1 | XP_023500803.1 | 3416 | 116 | |||
| XM_023645041.1 | XP_023500809.1 | 3290 | 116 | |||
| XM_023645050.1 | XP_023500818.1 | 3281 | 116 | |||
| XM_023645053.1 | XP_023500821.1 | 3221 | 116 | |||
| XM_023645058.1 | XP_023500826.1 | 3077 | 116 | |||
| XM_023645060.1 | XP_023500828.1 | 3236 | 116 | |||
Set Membership: |
|