Search
Search

AP3M1| Gene Symbol: AP3M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503944.6 | XP_001503994.1 | 964 | 111 | ||
| XM_005602499.3 | XP_005602556.1 | 1015 | 111 | |||
| XM_005602500.3 | XP_005602557.1 | 1042 | 111 | |||
| XM_014733028.2 | XP_014588514.1 | 891 | 111 | |||
| XM_023625238.1 | XP_023481006.1 | 1142 | 111 | |||
| GenBank mRNA | JL641205.1 | - | 1 - 2 | 145 | 111 | |
Set Membership: |
|