Search
Search
| Gene Symbol: HYKK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001492632.4 | XP_001492682.1 | 601 | 64 | ||
| XM_005602878.3 | XP_005602935.1 | 638 | 64 | |||
| XM_005602879.3 | XP_005602936.2 | 639 | 64 | |||
| XM_005602880.3 | XP_005602937.1 | 508 | 64 | |||
Set Membership: |
|