Search
Search

ZWILCH| Gene Symbol: ZWILCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023654987.1 | XP_023510755.1 | 1576 | 102 | ||
| XM_023654988.1 | XP_023510756.1 | 1583 | 102 | |||
| XM_023654991.1 | XP_023510759.1 | 1580 | 102 | |||
| XM_023654992.1 | XP_023510760.1 | 1575 | 102 | |||
| XM_023654993.1 | XP_023510761.1 | 1575 | 102 | |||
| XM_023654995.1 | XP_023510763.1 | 3043 | 102 | |||
| XM_023654998.1 | XP_023510766.1 | 1523 | 102 | |||
| GenBank mRNA | JL620705.1 | - | 10 - 11 | 925 | 102 | |
Set Membership: |
|