Search
Search

DUOXA1| Gene Symbol: DUOXA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014733954.2 | XP_014589440.1 | 722 | 107 | ||
| XM_014733955.2 | XP_014589441.1 | 659 | 107 | |||
| XM_023618369.1 | XP_023474137.1 | 402 | 107 | |||
| XM_023618370.1 | XP_023474138.1 | 880 | 107 | |||
| XM_023618371.1 | XP_023474139.1 | 792 | 107 | |||
| XM_023618375.1 | XP_023474143.1 | 723 | 107 | |||
| XM_023618378.1 | XP_023474146.1 | 465 | 107 | |||
| XM_023618379.1 | XP_023474147.1 | 810 | 107 | |||
| XM_023618385.1 | XP_023474153.1 | 653 | 107 | |||
| XM_023618397.1 | XP_023474165.1 | 741 | 107 | |||
| XM_023618398.1 | XP_023474166.1 | 747 | 107 | |||
| XM_023618415.1 | XP_023474183.1 | 590 | 107 | |||
Set Membership: |
|