Search
Search
| Gene Symbol: KNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014734033.2 | XP_014589519.1 | 6647 | 72 | ||
| XM_023619882.1 | XP_023475650.1 | 6692 | 72 | |||
| XM_023619890.1 | XP_023475658.1 | 6590 | 72 | |||
| XM_023619892.1 | XP_023475660.1 | 6641 | 72 | |||
| XM_023619904.1 | XP_023475672.1 | 6321 | 72 | |||
| XM_023619909.1 | XP_023475677.1 | 6315 | 72 | |||
| XM_023619916.1 | XP_023475684.1 | 6287 | 72 | |||