Search
Search

SUPT5H| Gene Symbol: SUPT5H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362312.4 | XP_003362360.1 | 1965 | 72 | ||
| XM_005596233.3 | XP_005596290.1 | 1977 | 72 | |||
| XM_014735351.2 | XP_014590837.1 | 1904 | 72 | |||