Search
Search

ABCD3| Gene Symbol: ABCD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005610406.3 | XP_005610463.1 | 1426 | 98 | ||
| XM_023641598.1 | XP_023497366.1 | 1563 | 98 | |||
| XM_023641599.1 | XP_023497367.1 | 1566 | 98 | |||
Set Membership: |
|