Search
Search

ARFGEF3| Gene Symbol: ARFGEF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023651189.1 | XP_023506957.1 | 331 | 88 | ||
| XM_023651190.1 | XP_023506958.1 | 213 | 88 | |||
| XM_023651191.1 | XP_023506959.1 | 331 | 88 | |||
| XM_023651192.1 | XP_023506960.1 | 331 | 88 | |||
| XM_023651193.1 | XP_023506961.1 | 306 | 88 | |||
| XM_023651194.1 | XP_023506962.1 | 331 | 88 | |||
Set Membership: |
|