Search
Search
| Gene Symbol: DRC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005597832.3 | XP_005597889.2 | 959 | 78 | ||
| XM_005597833.3 | XP_005597890.2 | 1006 | 78 | |||
| XM_005597834.3 | XP_005597891.2 | 793 | 78 | |||
| XM_014729107.2 | XP_014584593.2 | 676 | 78 | |||
| XM_023653656.1 | XP_023509424.1 | 1025 | 78 | |||
| XM_023653658.1 | XP_023509426.1 | 908 | 78 | |||
| XM_023653659.1 | XP_023509427.1 | 652 | 78 | |||
Set Membership: |
|