Search
Search
| Gene Symbol: CD82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001489923.6 | XP_001489973.4 | 504 | 109 | ||
| XM_014729435.2 | XP_014584921.2 | 457 | 109 | |||
| XM_023653917.1 | XP_023509685.1 | 473 | 109 | |||
| XM_023653918.1 | XP_023509686.1 | 378 | 109 | |||
| XM_023653919.1 | XP_023509687.1 | 360 | 109 | |||
Set Membership: |
|