Search
Search

FBXO38| Gene Symbol: FBXO38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503809.5 | XP_001503859.1 | 2956 | 126 | ||
| XM_003362788.4 | XP_003362836.1 | 2885 | 126 | |||
| XM_005599314.3 | XP_005599371.1 | 3110 | 126 | |||
| XM_005599315.3 | XP_005599372.1 | 3001 | 126 | |||
| XM_005599316.3 | XP_005599373.1 | 3003 | 126 | |||
| XM_005599317.3 | XP_005599374.1 | 3045 | 126 | |||
| XM_005599318.3 | XP_005599375.1 | 2374 | 126 | |||
| GenBank mRNA | JL640433.1 | - | 2 - 3 | 185 | 126 | |
Set Membership: |
|