Search
Search
| Gene Symbol: TLR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023632733.1 | XP_023488501.1 | 1087 | 85 | ||
| XM_023632734.1 | XP_023488502.1 | 747 | 85 | |||
| XM_023632735.1 | XP_023488503.1 | 228 | 85 | |||
| XM_023632736.1 | XP_023488504.1 | 953 | 85 | |||
Set Membership: |
|