Search
Search

PADI6| Gene Symbol: PADI6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023635646.1 | XP_023491414.1 | 4227 | 86 | ||
| XM_023635647.1 | XP_023491415.1 | 4120 | 86 | |||
| XM_023635648.1 | XP_023491416.1 | 4229 | 86 | |||
| XM_023635649.1 | XP_023491417.1 | 2722 | 86 | |||