Search
Search

FRMPD2| Gene Symbol: FRMPD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023648375.1 | XP_023504143.1 | 1443 | 67 | ||
| XM_023648382.1 | XP_023504150.1 | 1444 | 67 | |||
| XM_023648388.1 | XP_023504156.1 | 1444 | 67 | |||
| XM_023648392.1 | XP_023504160.1 | 989 | 67 | |||
| XM_023648398.1 | XP_023504166.1 | 4365 | 67 | |||
| XM_023648408.1 | XP_023504176.1 | 1159 | 67 | |||
| XM_023648418.1 | XP_023504186.1 | 1442 | 67 | |||
Set Membership: |
|