Search
Search

LOC102149641| Gene Symbol: LOC102149641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023650096.1 | XP_023505864.1 | 2565 | 64 | ||
| XM_023650097.1 | XP_023505865.1 | 2484 | 64 | |||
| XM_023650098.1 | XP_023505866.1 | 2459 | 64 | |||
| XM_023650099.1 | XP_023505867.1 | 3479 | 64 | |||
| XM_023650100.1 | XP_023505868.1 | 3440 | 64 | |||
| XM_023650101.1 | XP_023505869.1 | 2424 | 64 | |||
| XM_023650102.1 | XP_023505870.1 | 2421 | 64 | |||
| XM_023650103.1 | XP_023505871.1 | 2406 | 64 | |||
| XM_023650104.1 | XP_023505872.1 | 2406 | 64 | |||
| XM_023650105.1 | XP_023505873.1 | 2265 | 64 | |||
| XM_023650107.1 | XP_023505875.1 | 2262 | 64 | |||
| XM_023650108.1 | XP_023505876.1 | 2259 | 64 | |||
| XM_023650109.1 | XP_023505877.1 | 2247 | 64 | |||
| XM_023650111.1 | XP_023505879.1 | 1637 | 64 | |||
| XM_023650113.1 | XP_023505881.1 | 1473 | 64 | |||
Set Membership: |
|