Search
Search

FAM160B1| Gene Symbol: FAM160B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288933.2 | XP_015144419.1 | 4 - 5 | 627 | 66 | |
| XM_025151957.1 | XP_025007725.1 | 4 - 5 | 629 | 66 | ||
| XM_421774.6 | XP_421774.3 | 4 - 5 | 625 | 66 | ||
| GenBank mRNA | CR391114.1 | - | 2 - 3 | 202 | 66 | |
Set Membership: |
|