Search
Search

CEP192| Gene Symbol: CEP192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015282473.2 | XP_015137959.1 | 31 - 32 | 6127 | 71 | |
| XM_015282474.2 | XP_015137960.1 | 31 - 32 | 6248 | 71 | ||
| XM_015282475.2 | XP_015137961.1 | 31 - 32 | 6121 | 71 | ||
| XM_025147809.1 | XP_025003577.1 | 31 - 32 | 6245 | 71 | ||
| XM_025147810.1 | XP_025003578.1 | 31 - 32 | 6250 | 71 | ||
| XM_025147811.1 | XP_025003579.1 | 31 - 32 | 6250 | 71 | ||
| GenBank mRNA | CR391317.1 | - | 2 - 3 | 317 | 71 | |
Set Membership: |
|