Search
Search

PXDN| Gene Symbol: PXDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004940497.3 | XP_004940554.2 | 7 - 8 | 904 | 133 | |
| XM_419931.6 | XP_419931.4 | 8 - 9 | 977 | 133 | ||
| GenBank mRNA | CR523669.1 | - | 6 - 7 | 519 | 133 | |
Set Membership: |
|