Search
Search

| Gene Symbol: AAED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003643010.4 | XP_003643058.1 | 4 - 5 | 2947 | 75 | |
| GenBank mRNA | BX932659.1 | - | 3 - 4 | 390 | 75 | |
| Gene Symbol: CDC14B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025144748.1 | XP_025000516.1 | 1 - 2 | 1188 | 75 | |
Set Membership: |
|