Search
Search

IFI35| Gene Symbol: IFI35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418132.6 | XP_418132.1 | 3 - 4 | 373 | 74 | |
| GenBank mRNA | BX933445.2 | - | 3 - 4 | 330 | 74 | |
| BX934680.1 | - | 3 - 4 | 353 | 74 | ||
| BX934849.1 | - | 3 - 4 | 333 | 74 | ||
| KY931454.1 | - | 2 - 3 | 268 | 74 | ||
Set Membership: |
|