Search
Search

CUL5| Gene Symbol: CUL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015279528.2 | XP_015135014.1 | 2 - 3 | 478 | 65 | |
| XM_015279534.2 | XP_015135020.1 | 2 - 3 | 478 | 65 | ||
| GenBank mRNA | CR387212.1 | - | 1 - 2 | 79 | 65 | |
Set Membership: |
|