Search
Search

TICRR| Gene Symbol: TICRR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285869.1 | NP_001272798.1 | 9 - 10 | 2196 | 78 | |
| XM_025153845.1 | XP_025009613.1 | 9 - 10 | 2273 | 78 | ||
| GenBank mRNA | AM069775.1 | - | 2 - 3 | 182 | 78 | |
| BU208425.1 | - | 5 - 6 | 619 | 77 | ||
| CR407392.1 | - | 5 - 6 | 717 | 78 | ||
Set Membership: |
|