Search
Search

CCDC12| Gene Symbol: CCDC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006353.1 | NP_001006353.1 | 3 - 4 | 297 | 76 | |
| XM_025147149.1 | XP_025002917.1 | 3 - 4 | 253 | 76 | ||
| XM_025147150.1 | XP_025002918.1 | 2 - 3 | 290 | 76 | ||
| GenBank mRNA | AJ720250.1 | - | 3 - 4 | 297 | 76 | |
| CR523832.1 | - | 3 - 4 | 305 | 76 | ||
Set Membership: |
|