Search
Search

LAMP2| Gene Symbol: LAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001749.1 | NP_001001749.1 | 7 - 8 | 1010 | 72 | |
| XM_015278204.2 | XP_015133690.1 | 7 - 8 | 1051 | 72 | ||
| XM_015278205.2 | XP_015133691.1 | 7 - 8 | 1051 | 72 | ||
| GenBank mRNA | U10546.1 | - | 4 - 5 | 446 | 72 | |
| U10547.1 | - | 7 - 8 | 1010 | 72 | ||
| U10548.1 | - | 2 - 3 | 212 | 72 | ||
Set Membership: |
|