Search
Search

PTCH1| Gene Symbol: PTCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204960.2 | NP_990291.2 | 6 - 7 | 960 | 65 | |
| XM_015280248.2 | XP_015135734.1 | 6 - 7 | 929 | 65 | ||
| XM_025144670.1 | XP_025000438.1 | 5 - 6 | 607 | 65 | ||
| GenBank mRNA | U40074.1 | - | 6 - 7 | 957 | 65 | |
Set Membership: |
|