Search
Search

UGGT2| Gene Symbol: UGGT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252099.2 | NP_001239028.1 | 3 - 4 | 434 | 65 | |
| XM_015274455.2 | XP_015129941.1 | 3 - 4 | 427 | 65 | ||
| GenBank mRNA | AJ851758.1 | - | 3 - 4 | 435 | 65 | |
Set Membership: |
|