Search
Search

TXN2| Gene Symbol: TXN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031410.1 | NP_001026581.1 | 2 - 3 | 247 | 82 | |
| XM_015285949.2 | XP_015141435.1 | 2 - 3 | 255 | 82 | ||
| XM_025151067.1 | XP_025006835.1 | 2 - 3 | 2254 | 82 | ||
| GenBank mRNA | AJ720298.1 | - | 2 - 3 | 247 | 82 | |
| CR387816.1 | - | 1 - 2 | 218 | 82 | ||
| CR390224.1 | - | 1 - 2 | 229 | 82 | ||
Set Membership: |
|