Search
Search

CRHR2| Gene Symbol: CRHR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204454.1 | NP_989785.1 | 3 - 4 | 318 | 117 | |
| XM_015281045.2 | XP_015136531.1 | 4 - 5 | 475 | 117 | ||
| GenBank mRNA | AJ557031.2 | - | 3 - 4 | 318 | 117 | |
Set Membership: |
|