Search
Search

BFSP1| Gene Symbol: BFSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205151.1 | NP_990482.1 | 2 - 3 | 658 | 112 | |
| XM_025148550.1 | XP_025004318.1 | 9 - 10 | 3197 | 112 | ||
| XM_025148551.1 | XP_025004319.1 | 8 - 9 | 3087 | 112 | ||
| GenBank mRNA | X72873.1 | - | 658 | 112 | ||
Set Membership: |
|