Search
Search

HAO1| Gene Symbol: HAO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199442.2 | NP_001186371.2 | 5 - 6 | 817 | 66 | |
| XM_015283479.2 | XP_015138965.1 | 5 - 6 | 1152 | 66 | ||
| XM_015283480.2 | XP_015138966.1 | 5 - 6 | 2142 | 66 | ||
| XM_025148596.1 | XP_025004364.1 | 5 - 6 | 2182 | 66 | ||
Set Membership: |
|