Search
Search

CTSH| Gene Symbol: CTSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318407.1 | NP_001305336.1 | 1 - 2 | 75 | 97 | |
| GenBank mRNA | CR353641.1 | - | 1 - 2 | 99 | 97 | |
| JF514547.1 | - | 1 - 2 | 75 | 97 | ||
Set Membership: |
|