Search
Search

ASB15| Gene Symbol: ASB15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004937524.2 | XP_004937581.1 | 11 | 2315 | 109 | |
| XM_415996.4 | XP_415996.2 | 10 | 2268 | 109 | ||
| GenBank mRNA | CR338956.2 | - | 1 | 414 | 109 | |
Set Membership: |
|