Search
Search

SHPRH| Gene Symbol: SHPRH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935583.3 | XP_004935640.2 | 24 - 25 | 4569 | 161 | |
| XM_004935584.3 | XP_004935641.2 | 24 - 25 | 4566 | 161 | ||
Set Membership: |
|