Search
Search

COQ2| Gene Symbol: COQ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004941124.3 | XP_004941181.3 | 2 - 3 | 1107 | 110 | |
| XM_004941125.3 | XP_004941182.3 | 2 - 3 | 1171 | 110 | ||
| XM_420564.6 | XP_420564.5 | 2 - 3 | 1061 | 110 | ||
Set Membership: |
|