Search
Search

KLHL8| Gene Symbol: KLHL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015276505.2 | XP_015131991.1 | 10 - 11 | 2251 | 79 | |
| XM_015276506.2 | XP_015131992.1 | 9 - 10 | 1986 | 79 | ||
| XM_025150378.1 | XP_025006146.1 | 11 - 12 | 3633 | 79 | ||
| XM_025150379.1 | XP_025006147.1 | 11 - 12 | 4112 | 79 | ||
Set Membership: |
|