Search
Search

PTPN7| Gene Symbol: PTPN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298763.2 | XP_015154249.1 | 676 | 158 | ||
| XM_015298765.2 | XP_015154251.1 | 792 | 158 | |||
| XM_015298766.2 | XP_015154252.1 | 2413 | 158 | |||
Set Membership: |
|