Search
Search

ARMC9| Gene Symbol: ARMC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004943365.3 | XP_004943422.1 | 2 - 3 | 349 | 83 | |
| XM_025153644.1 | XP_025009412.1 | 2 - 3 | 352 | 83 | ||
| XM_025153645.1 | XP_025009413.1 | 2 - 3 | 351 | 83 | ||
| XM_025153646.1 | XP_025009414.1 | 2 - 3 | 357 | 83 | ||
| XM_025153647.1 | XP_025009415.1 | 2 - 3 | 396 | 83 | ||
| XM_025153648.1 | XP_025009416.1 | 2 - 3 | 396 | 83 | ||
| GenBank mRNA | BX934271.1 | - | 2 - 3 | 350 | 83 | |
Set Membership: |
|