Search
Search

LOC100785449| Gene Symbol: LOC100785449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252921.2 | NP_001239850.1 | 4 - 5 | 709 | 83 | |
| GenBank mRNA | BT095021.1 | - | 1 - 2 | 476 | 83 | |
Set Membership: |
|