Search
Search

LOC100819292
LOC100812344| Gene Symbol: LOC100819292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK244394.1 | - | 3 - 4 | 731 | 95 | |
| AK244395.1 | - | 3 - 4 | 731 | 95 | ||
| Gene Symbol: LOC100812344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK243915.1 | - | 3 - 4 | 740 | 95 | |
Set Membership: |
|