Search
Search

LOC100805924| Gene Symbol: LOC100805924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252890.2 | NP_001239819.1 | 8 | 1216 | 97 | |
| GenBank mRNA | BT099497.1 | - | 1 | 132 | 97 | |
Set Membership: |
|