Search
Search

Aig1| Gene Symbol: Aig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846046.2 | XP_004846103.1 | 1 - 2 | 238 | 69 | |
| XM_021259321.1 | XP_021114980.1 | 1 - 2 | 240 | 69 | ||
| XM_021259323.1 | XP_021114982.1 | 1 - 2 | 240 | 69 | ||
Set Membership: |
|