Search
Search

Fank1| Gene Symbol: Fank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021253288.1 | XP_021108947.1 | 7 - 8 | 779 | 64 | |
| XM_021253289.1 | XP_021108948.1 | 6 - 7 | 713 | 64 | ||
| XM_021253290.1 | XP_021108949.1 | 7 - 8 | 865 | 64 | ||
Set Membership: |
|