Search
Search

Banp| Gene Symbol: Banp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842696.2 | XP_004842753.1 | 2 - 3 | 1139 | 62 | |
| XM_004842697.2 | XP_004842754.1 | 2 - 3 | 1352 | 62 | ||
| XM_013071446.2 | XP_012926900.1 | 2 - 3 | 1355 | 62 | ||
| XM_013071447.2 | XP_012926901.1 | 2 - 3 | 1355 | 62 | ||
| XM_013071448.2 | XP_012926902.1 | 2 - 3 | 1140 | 62 | ||
| XM_013071449.2 | XP_012926903.1 | 2 - 3 | 1352 | 62 | ||
| XM_021256586.1 | XP_021112245.1 | 2 - 3 | 159 | 62 | ||
| XM_021256587.1 | XP_021112246.1 | 2 - 3 | 1370 | 62 | ||
Set Membership: |
|