Search
Search

Nr1h2| Gene Symbol: Nr1h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866827.3 | XP_004866884.1 | 7 - 8 | 1379 | 61 | |
| XM_004866828.3 | XP_004866885.1 | 7 - 8 | 1238 | 61 | ||
| XM_021245810.1 | XP_021101469.1 | 7 - 8 | 1382 | 61 | ||
| XM_021245811.1 | XP_021101470.1 | 6 - 7 | 1344 | 61 | ||
| XM_021245812.1 | XP_021101471.1 | 7 - 8 | 1241 | 61 | ||
| XM_021245813.1 | XP_021101472.1 | 6 - 7 | 1109 | 61 | ||
| XM_021245814.1 | XP_021101473.1 | 5 - 6 | 1074 | 61 | ||
| XM_021245815.1 | XP_021101474.1 | 6 - 7 | 941 | 61 | ||
Set Membership: |
|