Search
Search

Nudcd3| Gene Symbol: Nudcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839911.3 | XP_004839968.1 | 3 - 4 | 834 | 69 | |
| XM_021254279.1 | XP_021109938.1 | 3 - 4 | 840 | 69 | ||
Set Membership: |
|