Search
Search

Rabl6| Gene Symbol: Rabl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848849.3 | XP_004848906.1 | 8 - 9 | 949 | 55 | |
| XM_021261655.1 | XP_021117314.1 | 6 - 7 | 762 | 55 | ||
| XM_021261656.1 | XP_021117315.1 | 9 - 10 | 1866 | 55 | ||
| XM_021261657.1 | XP_021117316.1 | 8 - 9 | 1771 | 55 | ||
Set Membership: |
|